Lead(II) acetate trihydrate 99.0-103.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00150023
Beilstein Registry No.: 3730298
Notes: pH of a 5% aqueous solution at 25°C = 5.5-6.5, Decomposes above 200°C
Freely soluble in water, alcohol, and glycerol
Purity: 99.0-103.0%
CAS No.: 6080-56-4
Molecular Formula: Pb(OOCCH3)2.3H2O
Formula Weight: 379.33 (325.28anhy)
Physical Form: Granular
Density: 2.55
Melting Point: 75°
Solubility: Freely soluble in water, alcohol, and glycerol
Beilstein Reference: 3730298
Merck Reference: 14,5397
UN No.: 1616
MDL No.: MFCD00150023
Formula:
(H₃CCOO)₂Pb·3H₂O Boiling Pt: 280 °C (1013 hPa) Melting Pt: 75 °C Density: 2.55 Storage Temperature: Ambient |
MDL Number:
MFCD00150023 CAS Number: 6080-56-4 EINECS: 206-104-4 UN: 1616 ADR: 6.1,III Merck Index: 14,5397 |
|
Specification Test Results
Insoluble matter | 0.01% |
Cl | 5 ppm |
Nitrate and nitrite (as NO3) | 0.005% |
Ca | 0.005% |
Cu | 0.002% |
K | 0.005% |
NA | 0.0001 |
Fe | 0.00001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...