Sodium selenate decahydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14239-14
14239-22
CAAA14239-14EA
170.64
CAD
CAAA14239-14
CAAA14239-22
Sodium selenate decahydrate ≥99.9% (metals basis)
Sodium selenate decahydrate
MDL: MFCD00149174
Soluble in water
Purity: 99.9%
CAS No.: 10102-23-5
Molecular Formula: Na2SeO4.10H2O
Formula Weight: 369.09 (188.94anhy)
Physical Form: Crystalline
Density: 1.600
Melting Point: 35° dec.
Solubility: Soluble in water
Merck Reference: 14,8671
UN No.: 2630
MDL No.: MFCD00149174
Formula:
Na₂SeO₄·10H₂O Melting Pt: 35 °C (dec.) Density: 1.600 Storage Temperature: Ambient |
MDL Number:
MFCD00149174 CAS Number: 10102-23-5 EINECS: 236-501-8 UN: 2630 ADR: 6.1,I Merck Index: 14,8671 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...