Cesium hydrogen carbonate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

14234-14 14234-22
CAAA14234-14EA 161.16 CAD
CAAA14234-14 CAAA14234-22
Cesium hydrogen carbonate ≥99.99% (metals basis)
Cesium hydrogen carbonate

MDL: MFCD00010961


Purity: 99.99% CAS No.: 29703-01-3 Molecular Formula: CsHCO3 Formula Weight: 193.92 Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00010961

Formula: CHCsO₃
Melting Pt: 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010961
CAS Number: 15519-28-5
EINECS: 249-784-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR