Aluminium ≥99% (metals basis), foil [Si & Fe (combined) ∼ 1%], annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

14196-CJ 14196-RW 14196-RX
CAAA14196-CJEA 151.68 CAD
CAAA14196-CJ CAAA14196-RW CAAA14196-RX
Aluminium ≥99% (metals basis), foil [Si & Fe (combined) ∼ 1%], annealed, Thickness: 0.25 mm (0.01 in)
Aluminium

MDL: MFCD00134029


Purity: 99% CAS No.: 7429-90-5 Physical Form: ≈102.8g/30x50cm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR