Platinum ≥99.95% (metals basis), wire, hard, Ø 0.762 mm (0.03 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Platinium
14195-BV
14195-BW
14195-BW
14195-BS
14195-BS
14195-BV
CAAA14195-BVEA
1211.86
CAD
AA14195-BW
CAAA14195-BV
AA14195-BS
CAAA14195-BW
AA14195-BV
CAAA14195-BS
Platinum ≥99.95% (metals basis), wire, hard, Ø 0.762 mm (0.03 in)
Platinum
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 °C (1013 hPa) Melting Pt: 1769 °C Density: 21.447 g/cm³ (20 °C) |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...