Nickel ≥99.5% (metals basis), wire, annealed, Ø 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14188-G5
14188-G9
CAAA14188-G5EA
169.06
CAD
CAAA14188-G5
CAAA14188-G9
Nickel ≥99.5% (metals basis), wire, annealed, Ø 2.0 mm (0.08 in)
Nickel
MDL: MFCD00011137
Purity: 99.5%
CAS No.: 7440-02-0
Physical Form: ≈28.0g/m, Temper: soft
Odor: Odorless
MDL No.: MFCD00011137
Formula:
Ni Boiling Pt: 3007 °C (1013 hPa) Melting Pt: 1453 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...