Iron ≥99% (metals basis), powder -20 mesh

Supplier: Thermo Scientific Chemicals
Warning

14183-A1 14183-A7
CAAA14183-A1EA 429.76 CAD
CAAA14183-A1 CAAA14183-A7
Iron ≥99% (metals basis), powder -20 mesh
Iron

MDL: MFCD00010999


Purity: 99% CAS No.: 7439-89-6 MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1536 °C
Density: 7,86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR