Copper(II) sulfate pentahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
14178-36
14178-A3
CAAA14178-36EA
121.5
CAD
CAAA14178-36
CAAA14178-A3
Copper(II) sulfate pentahydrate 98.0-102.0% ACS
Copper (II) sulphate pentahydrate
MDL: MFCD00149681
Notes: m.p. 30°C -2H2O, 110°C -4H2O, 250°C anhydrous; ñ 2.286
Soluble in water, methanol, glycerol.
Fieser: 1,164 2,89 5,162 6,141 10,107 19,111
Purity: 98.0-102.0%
CAS No.: 7758-99-8
Molecular Formula: CuSO4.5H2O
Formula Weight: 249.68 (159.60anhy)
Physical Form: Crystalline
Density: 2.270
Appearance: Blue
Odor: Odorless
pH: 4
Solubility: Soluble in water, methanol, glycerol.
Merck Reference: 14,2653
UN No.: 3288
MDL No.: MFCD00149681
Formula:
CuSO₄·5H₂O Density: 2.270 Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3288 ADR: 6.1,III REACH: 01-2119520566-40 Merck Index: 14,2653 |
Specification Test Results
Insoluble matter | 0.005% |
Cl | 0.001% |
Nitrogen compounds (as N) | 0.002% |
Ca | 0.005% |
Ni | 0.005% |
K | 0.01% |
NA | 0.0002 |
Fe | 0.00003 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...