Copper(II) acetate monohydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

14177-22 14177-36 14177-A3
CAAA14177-22EA 87.06 CAD
CAAA14177-22 CAAA14177-36 CAAA14177-A3
Copper(II) acetate monohydrate 98.0-102.0% ACS
Copper (II) acetate monohydrate

MDL: MFCD00149570


Purity: 98.0-102.0% CAS No.: 6046-93-1 Molecular Formula: C4H6CuO4.H2O Formula Weight: 199.65 (181.64anhy) Physical Form: Powder Density: 1.882 Appearance: Blue-green Odor: Odorless pH: 5.2-5.5 Melting Point: 115° Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00149570

Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115 °C
Density: 1.882
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9 III
Merck Index: 14,2624

Order Now

Specification Test Results

Insoluble matter 0.01
Cl 0.003
SO3 0.01
Ca 0.005%
K 0.01%
NA 0.05%
Fe 0.002%
Ni 0.0001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR