Rhenium ≥99.97% (metals basis), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
14169-BP
14169-BS
14169-BW
CAAA14169-BPEA
127.98
CAD
CAAA14169-BP
CAAA14169-BS
CAAA14169-BW
Rhenium ≥99.97% (metals basis), wire, Ø 1.0 mm (0.04 in)
Rhenium
MDL: MFCD00011195
Purity: 99.97%
CAS No.: 7440-15-5
Physical Form: ≈16.52g/m
Odor: Odorless
MDL No.: MFCD00011195
Formula:
Re Boiling Pt: 5630 °C (1013 hPa) Melting Pt: 3180 °C Density: 21.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 Merck Index: 12,08343 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...