Silicon carbide, β-phase ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

14165-22 14165-36 14165-A3
CAAA14165-22EA 151.68 CAD
CAAA14165-22 CAAA14165-36 CAAA14165-A3
Silicon carbide, β-phase ≥99.8% (metals basis)
Silicon carbide, β-phase

MDL: MFCD00049531


Purity: 99.8% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 1 Micron Powder, S.A. 14-19m2/g Odor: Odorless Merck Reference: 14,8492 MDL No.: MFCD00049531

Formula: SiC
MW: 40.10 g/mol
Melting Pt: 2700 °C
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR