Zinc sulfate monohydrate Zn 35.5%

Supplier: Thermo Scientific Chemicals
Danger

14160-A1 14160-A7
CAAA14160-A1EA 113.29 CAD
CAAA14160-A1 CAAA14160-A7
Zinc sulfate monohydrate Zn 35.5%
Zinc sulphate monohydrate

MDL: MFCD00149893 Notes: greater than 238°C -H2O Soluble in water


CAS No.: 7446-19-7 Molecular Formula: ZnSO4.H2O Formula Weight: 179.45 (161.43anhy) Physical Form: Coarse Powder Solubility: Soluble in water Merck Reference: 14,10159 MDL No.: MFCD00149893

Formula: ZnSO₄·1H₂O
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 14,10159

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR