Silver ≥99.999% (metals basis), shot, Premion® 2-3 mm (0.08-0.12 in)
Supplier: Thermo Scientific Chemicals
14153-09
14153-18
14153-30
CAAA14153-09EA
219.62
CAD
CAAA14153-09
CAAA14153-18
CAAA14153-30
Silver ≥99.999% (metals basis), shot, Premion® 2-3 mm (0.08-0.12 in)
Silver
For vapor deposition
Purity: 99.999%
CAS No.: 7440-22-4
Odor: Characteristic
MDL No.: MFCD00003397
For vapor deposition
Formula:
Ag Boiling Pt: 2212 °C (1013 hPa) Melting Pt: 962 °C Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...