Tin ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
14148-BU
14148-BY
CAAA14148-BUEA
169.06
CAD
CAAA14148-BU
CAAA14148-BY
Tin ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Tin
MDL: MFCD00133862
Purity: 99.9%
CAS No.: 7440-31-5
Physical Form: ≈9.23g/cm
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...