Nickel(II) fluoride tetrahydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

14132-18 14132-30
CAAA14132-18EA 74.42 CAD
CAAA14132-18 CAAA14132-30
Nickel(II) fluoride tetrahydrate 98+%
Nickel (II) fluoride tetrahydrate

MDL: MFCD00016263


Purity: 98+% CAS No.: 13940-83-5 Molecular Formula: NiF2.4H2O Formula Weight: 168.77 (96.71anhy) Physical Form: Powder Odor: Odorless UN No.: 3288 MDL No.: MFCD00016263

Formula: F₂Ni·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00016263
CAS Number: 13940-83-5
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR