Dibutyltin oxide

Supplier: Thermo Scientific Chemicals
Danger

14117-18 14117-A1
CAAA14117-18EA 38.24 CAD
CAAA14117-18 CAAA14117-A1
Dibutyltin oxide
Dibutyltin oxide

MDL: MFCD00001992 Beilstein Registry No.: 4126243 Fieser: 5,188 10,123 12,160 13,95 15,116 16,112 18,125 20,133 21,163


CAS No.: 818-08-6 Molecular Formula: C8H18OSn Formula Weight: 248.92 Physical Form: Powder Density: 1.58 Appearance: White Odor: Odorless Melting Point: 300° Beilstein Reference: 4126243 UN No.: 3146 MDL No.: MFCD00001992

Formula: C₈H₁₈OSn
Melting Pt: >300 °C
Density: 1,58
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR