Dibutyltin dichloride ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14116-09
14116-30
14116-A1
CAAA14116-09EA
43.13
CAD
CAAA14116-09
CAAA14116-30
CAAA14116-A1
Dibutyltin dichloride ≥96%
Dibutyltin dichloride
Organotin intermediate
Purity: 96%
CAS No.: 683-18-1
Molecular Formula: C8H18Cl2Sn
Formula Weight: 303.83
Flash Point: 168°(334°F)
Physical Form: Crystalline
Density: 1.36
Refractive Index: 1.4991
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Acrid
Melting Point: 39-41°
Boiling Point: 135°/10mm
Solubility: Insoluble in cold water. Hydrolyzed by hot water. Soluble in organic solvents
UN No.: 2928
MDL No.: MFCD00000518
Organotin intermediate
Formula:
C₈H₁₈Cl₂Sn Boiling Pt: 135 °C (10 mmHg) Melting Pt: 39…41 °C Density: 1,36 Flash Pt: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000518 CAS Number: 683-18-1 EINECS: 211-670-0 UN: 2928 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...