Tetrabutyltin ≥95%

Supplier: Thermo Scientific Chemicals
Danger

14115-22 14115-36
CAAA14115-22EA 76 CAD
CAAA14115-22 CAAA14115-36
Tetrabutyltin ≥95%
Tetrabutyltin

MDL: MFCD00009422


Purity: 95% CAS No.: 1461-25-2 Molecular Formula: [CH3(CH2)3]4Sn Formula Weight: 347.17 Flash Point: 107°(225°F) Physical Form: Liquid Density: 1.057 Refractive Index: 1.474 Odor: Mild Melting Point: -97° Boiling Point: 144-146°/10mm UN No.: 2788 MDL No.: MFCD00009422

Formula: C₁₆H₃₆Sn
Boiling Pt: 144…146 °C (10 mmHg)
Melting Pt: –97 °C
Density: 1,057
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR