Tin(II) oxalate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tin (II) ethanedioate, Oxalic acid tin (II) salt
14113-22
14113-36
CAAA14113-22EA
56.72
CAD
CAAA14113-22
CAAA14113-36
Tin(II) oxalate
Tin (II) oxalate
MDL: MFCD00040678
Beilstein Registry No.: 3708588
Soluble in dilute HCl. Insoluble in water
CAS No.: 814-94-8
Molecular Formula: C2O4Sn
Formula Weight: 206.72
Physical Form: Powder
Density: 3.56
Appearance: White
Melting Point: 280° dec.
Solubility: Soluble in dilute HCl. Insoluble in water
Beilstein Reference: 3708588
Merck Reference: 14,8786
UN No.: 3288
MDL No.: MFCD00040678
Formula:
C₂O₄Sn MW: 206.73 g/mol Melting Pt: 280 °C (dec.) Density: 3.56 Storage Temperature: Ambient |
MDL Number:
MFCD00040678 CAS Number: 814-94-8 EINECS: 212-414-0 UN: 3288 ADR: 6.1,III Merck Index: 14,8786 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...