Triphenylphosphine 99+%

Supplier: Thermo Scientific Chemicals
Warning

14112-14 14112-22 14112-36
CAAA14112-14EA 53.56 CAD
CAAA14112-14 CAAA14112-22 CAAA14112-36
Triphenylphosphine 99+%
Triphenylphosphine

MDL: MFCD00003043 Beilstein Registry No.: 610776 Fieser: 1,1238 15,352 16,366 17,389 18,385 19,382 20,411 21,462


Purity: 99+% CAS No.: 603-35-0 Molecular Formula: (C6H5)3P Formula Weight: 262.29 Flash Point: 181°(358°F) Physical Form: Flake Density: 1.200 Appearance: White to pale cream Odor: Mild Melting Point: 79-81° Boiling Point: 360° Beilstein Reference: 610776 Merck Reference: 14,9743 MDL No.: MFCD00003043

Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377
Melting Pt: 79
Density: 1.200
Flash Pt: 181 °C (358 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 14,9743

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR