Sodium carbonate monohydrate ≥85%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium carbonate hydrous, Soda monohydrate
14108-A1
14108-A7
CAAA14108-A1EA
86.74
CAD
CAAA14108-A1
CAAA14108-A7
Sodium carbonate monohydrate ≥85%
Sodium carbonate monohydrate
MDL: MFCD00149177
Soluble in water. Insoluble in alcohol
Purity: 85% min
CAS No.: 5968-11-6
Molecular Formula: Na2CO3.H2O
Formula Weight: 124.01 (105.99anhy)
Physical Form: Crystalline
Density: 2.250
Appearance: Colorless to white
Odor: Odorless
Melting Point: 100° -H2O
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,8596
MDL No.: MFCD00149177
Formula:
Na₂CO₃·1H₂O Melting Pt: 100 °C (-H₂O) Density: 2.250 Storage Temperature: Ambient |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 EINECS: 207-838-8 Merck Index: 14,8596 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...