Disodium metasilicate pentahydrate, tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium metasilicate pentahydrate

14103-A1 14103-D9
CAAA14103-A1EA 84.21 CAD
CAAA14103-A1 CAAA14103-D9
Disodium metasilicate pentahydrate, tech.
di-Sodium metasilicate pentahydrate

MDL: MFCD00149176


CAS No.: 10213-79-3 Molecular Formula: Na2SiO3.5H2O Formula Weight: 212.14 (122.06anhy) Physical Form: -18 Mesh Granular Density: 1.75 Appearance: White Odor: Odorless Melting Point: 72° Merck Reference: 14,8642 UN No.: 3253 MDL No.: MFCD00149176

Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 72 °C
Density: 1.75
Storage Temperature: Ambient
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 14,8642

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR