Sodium bromate 99,5 Metals basis

Supplier: Thermo Scientific Chemicals
Danger

14101-22 14101-36 14101-A3
CAAA14101-22EA 55.46 CAD
CAAA14101-22 CAAA14101-36 CAAA14101-A3
Sodium bromate 99,5 Metals basis
Sodium bromate

MDL: MFCD00003476 Soluble in water (2.5 parts in cold water, 1.1 part in boiling water)


Purity: 99.5% CAS No.: 7789-38-0 Molecular Formula: NaBrO3 Formula Weight: 150.90 Physical Form: Crystalline Density: 3.34 Refractive Index: 1.594 Appearance: White Odor: Odorless Melting Point: 381° dec. Solubility: Soluble in water (2.5 parts in cold water, 1.1 part in boiling water) Merck Reference: 14,8593 UN No.: 1494 MDL No.: MFCD00003476

Formula: BrNaO₃
MW: 150.89 g/mol
Boiling Pt: 1390
Melting Pt: 381 (dec.)
Density: 3.34
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1 II
Merck Index: 14,8593

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR