Tetraethyl orthosilicate (TEOS) 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
As a binding agent and as a polymerization catalyst
Purity: 99+%
CAS No.: 78-10-4
Molecular Formula: C8H20O4Si
Formula Weight: 208.33
Flash Point: 46°(116°F)
Physical Form: Liquid
Density: 0.934
Refractive Index: 1.3820
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Pleasant
Melting Point: -85°
Boiling Point: 168°
Beilstein Reference: 1422225
Merck Reference: 14,3851
UN No.: 1292
MDL No.: MFCD00009062
As a binding agent and as a polymerization catalyst
Formula:
Si(OC₂H₅)₄ Boiling Pt: 168 °C Melting Pt: –85 °C Density: 0.934 Flash Pt: 46 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 14,3851 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...