Dichloro(methyl)silane 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyldichlorosilane

14079-22 14079-36
CAAA14079-22EA 85.79 CAD
CAAA14079-22 CAAA14079-36
Dichloro(methyl)silane 97+%
Dichloro(methyl)silane

MDL: MFCD00000494 Beilstein Registry No.: 1071194


Purity: 97+% CAS No.: 75-54-7 Molecular Formula: CH4Cl2Si Formula Weight: 115.03 Flash Point: -32°(-26°F) Physical Form: Liquid Density: 1.105 Refractive Index: 1.3980 Sensitivity: Air and Moisture Sensitive Appearance: Colorless Melting Point: -93° Boiling Point: 41-42° Beilstein Reference: 1071194 UN No.: 1242 MDL No.: MFCD00000494

Formula: CH₄Cl₂Si
MW: 115.03 g/mol
Boiling Pt: 41…42 °C
Melting Pt: –93 °C
Density: 1,105
Flash Pt: –32 °C (–26 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000494
CAS Number: 75-54-7
EINECS: 200-877-1
UN: 1242
ADR: 4.3,I
REACH: 01-2119436669-21

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR