Tantalum pentafluoride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14052-09
14052-18
CAAA14052-09EA
259.12
CAD
CAAA14052-09
CAAA14052-18
Tantalum pentafluoride ≥99.9% (metals basis)
Tantalum pentafluoride
MDL: MFCD00042542
Decomposes in water forming tantalum oxyfluoride. Soluble in conc. and fuming HNO3.
Purity: 99.9%
CAS No.: 7783-71-3
Molecular Formula: TaF5
Formula Weight: 275.94
Physical Form: Powder
Density: 4.74
Appearance: White
Melting Point: 96.8°
Boiling Point: 229.5°
Solubility: Decomposes in water forming tantalum oxyfluoride. Soluble in conc. and fuming HNO3
Merck Reference: 14,9055
UN No.: 3260
MDL No.: MFCD00042542
Formula:
F₅Ta Boiling Pt: 229.5 °C Melting Pt: 96.8 °C Density: 4.74 Storage Temperature: Ambient |
MDL Number:
MFCD00042542 CAS Number: 7783-71-3 EINECS: 232-022-3 UN: 3260 ADR: 8,II Merck Index: 14,9055 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...