Tin (II) sulfide ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14051-06
14051-14
CAAA14051-06EA
105.86
CAD
CAAA14051-06
CAAA14051-14
Tin (II) sulfide ≥99.5%
Tin (II) sulphide
Polymerization catalyst
Purity: 99.5%
CAS No.: 1314-95-0
Molecular Formula: SnS
Formula Weight: 150.75
Physical Form: -4 Mesh
Density: 5.08
Appearance: Grey to black
Melting Point: 882°
Solubility: Soluble in conc. HCl, hot conc. H2SO4
Merck Reference: 14,8791
MDL No.: MFCD00011245
Polymerization catalyst
Formula:
SSn Melting Pt: 882 °C Density: 5.08 Storage Temperature: Ambient |
MDL Number:
MFCD00011245 CAS Number: 1314-95-0 EINECS: 215-248-7 Merck Index: 14,8791 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...