Tantalum pentabromide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14041-09
14041-18
14041-30
CAAA14041-09EA
117.55
CAD
CAAA14041-09
CAAA14041-18
CAAA14041-30
Tantalum pentabromide ≥99.9% (metals basis)
Tantalum pentabromide
MDL: MFCD00049563
Purity: 99.9%
CAS No.: 13451-11-1
Molecular Formula: TaBr5
Formula Weight: 580.49
Physical Form: -8 Mesh Powder
Density: 4.67
Appearance: Yellow
Odor: Pungent
Melting Point: 265°
Boiling Point: 348.8°
UN No.: 3260
MDL No.: MFCD00049563
Formula:
Br₅Ta MW: 580.47 g/mol Boiling Pt: 348.8 °C Melting Pt: 265 °C Density: 4,67 Storage Temperature: Ambient |
MDL Number:
MFCD00049563 CAS Number: 13451-11-1 EINECS: 236-618-4 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...