Vanadium(III) chloride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
14039-09
14039-18
CAAA14039-09EA
167.48
CAD
CAAA14039-09
CAAA14039-18
Vanadium(III) chloride ≥99% (metals basis)
Vanadium (III) chloride
MDL: MFCD00011454
Purity: 99%
CAS No.: 7718-98-1
Molecular Formula: VCl3
Formula Weight: 157.30
Physical Form: -4 Mesh
Density: 3.00
Appearance: Pink to purple
Odor: Acrid
UN No.: 2475
MDL No.: MFCD00011454
Formula:
Cl₃V Density: 3,00 Storage Temperature: Ambient |
MDL Number:
MFCD00011454 CAS Number: 7718-98-1 EINECS: 231-744-6 UN: 2475 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...