Aluminium carbide 99+% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum carbide, Tetraaluminium tricarbide

14038-18 14038-30
CAAA14038-18EA 492.96 CAD
CAAA14038-18 CAAA14038-30
Aluminium carbide 99+% (metals basis)
Aluminium carbide

MDL: MFCD00003425 Notes: Decomposes in water with the evolution of methane


Purity: 99+% CAS No.: 1299-86-1 Molecular Formula: Al4C3 Formula Weight: 143.96 Physical Form: -325 Mesh, typically 10 microns or less Density: 2.36 Appearance: Dark grey Melting Point: 2100° (dec. 2200°) Merck Reference: 14,335 UN No.: 1394 MDL No.: MFCD00003425

Formula: C₃Al₄
MW: 143.96
Melting Pt: 2100
Density: 2.36
Storage Temperature: Ambient
MDL Number: MFCD00003425
CAS Number: 1299-86-1
EINECS: 215-076-2
UN: 1394
ADR: 4.3 II
Merck Index: 14,335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR