Sodium bromide 97%
Supplier: Thermo Scientific Chemicals
Warning
|
14037-36
14037-A3
CAAA14037-36EA
100.49
CAD
CAAA14037-36
CAAA14037-A3
Sodium bromide 97%
Sodium bromide
MDL: MFCD00003475
Soluble in water, alcohol
Fieser: 12,445
Purity: 97%
CAS No.: 7647-15-6
Molecular Formula: NaBr
Formula Weight: 102.89
Physical Form: Granular
Density: 3.203
Refractive Index: 1.6412
Sensitivity: Hygroscopic
Melting Point: 755°
Boiling Point: 1390°
Solubility: Soluble in water, alcohol
Merck Reference: 14,8594
MDL No.: MFCD00003475
Formula:
NaBr Boiling Pt: 1390 °C Melting Pt: 755 °C Density: 3.203 Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 Merck Index: 14,8594 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...