Molybdenum trichloride ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

14034-04 14034-09
CAAA14034-04EA 119.92 CAD
CAAA14034-04 CAAA14034-09
Molybdenum trichloride ≥99.5% (metals basis)
Molybdenum trichloride

MDL: MFCD00014214


Purity: 99.5% CAS No.: 13478-18-7 Molecular Formula: MoCl3 Formula Weight: 202.32 Physical Form: -100 Mesh Powder Sensitivity: Air and Moisture Sensitive Appearance: Dark red Odor: Acidic Melting Point: dec. UN No.: 3260 MDL No.: MFCD00014214

Formula: Cl₃Mo
Melting Pt: Dec.
Density: 3,58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014214
CAS Number: 13478-18-7
EINECS: 236-775-9
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR