Tetrabromosilane
Supplier: Thermo Scientific Chemicals
|
Danger
|
14029-09
14029-18
CAAA14029-09EA
152.63
CAD
CAAA14029-09
CAAA14029-18
Tetrabromosilane
Tetrabromosilane
MDL: MFCD00049530
CAS No.: 7789-66-4
Molecular Formula: SiBr4
Formula Weight: 347.72
Physical Form: Liquid
Density: 2.800
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Odor: Stench
Melting Point: 5°
Boiling Point: 154°
Merck Reference: 14,8499
UN No.: 3264
MDL No.: MFCD00049530
Formula:
Br₄Si Boiling Pt: 154 °C Melting Pt: 5 °C Density: 2.800 Storage Temperature: Ambient |
MDL Number:
MFCD00049530 CAS Number: 7789-66-4 EINECS: 232-182-4 UN: 3264 ADR: 8,II Merck Index: 14,8499 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...