Iron(II) sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

14024-03 14024-09 14024-18
CAAA14024-03EA 60.2 CAD
CAAA14024-03 CAAA14024-09 CAAA14024-18
Iron(II) sulfide ≥99.9% (metals basis)
Iron (II) sulphide

MDL: MFCD00011013


Purity: 99.9% CAS No.: 1317-37-9 Molecular Formula: FeS Formula Weight: 87.91 Physical Form: -100 Mesh Powder Density: 4.74 Appearance: Brown to black Odor: Odorless Melting Point: 1195° Merck Reference: 14,4058 MDL No.: MFCD00011013

Formula: FeS
Melting Pt: 1195 °C
Density: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 14,4058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR