Germanium(IV) iodide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetraiodogermane, Germanium tetraiodide

14009-09 14009-18
CAAA14009-09EA 398.16 CAD
CAAA14009-09 CAAA14009-18
Germanium(IV) iodide ≥99.999% (metals basis)
Germanium (IV) iodide

MDL: MFCD00016120


Purity: 99.999% CAS No.: 13450-95-8 Molecular Formula: GeI4 Formula Weight: 580.21 Physical Form: -10 Mesh Crystalline Density: 4.32 Melting Point: 146° UN No.: 3260 MDL No.: MFCD00016120

Formula: GeI₄
MW: 580.23
Boiling Pt: 350
Melting Pt: 146
Density: 4.32
Storage Temperature: Ambient
MDL Number: MFCD00016120
CAS Number: 13450-95-8
EINECS: 236-613-7
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR