Barium peroxide, anhydrous ≥84%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
14008-18
14008-30
14008-A1
14008-A7
CAAA14008-18EA
30.34
CAD
CAAA14008-18
CAAA14008-30
CAAA14008-A1
CAAA14008-A7
Barium peroxide, anhydrous ≥84%, Technical Grade
Barium peroxide
MDL: MFCD00003454
Insoluble in water.
Purity: 84% min
CAS No.: 1304-29-6
Molecular Formula: BaO2
Formula Weight: 169.34
Physical Form: Powder
Density: 4.96
Appearance: Tan
Melting Point: 450° dec.
Solubility: Insoluble in water
Merck Reference: 14,989
UN No.: 1449
MDL No.: MFCD00003454
Formula:
BaO₂ Melting Pt: 450 °C (dec.) Density: 4.96 Storage Temperature: Ambient |
MDL Number:
MFCD00003454 CAS Number: 1304-29-6 EINECS: 215-128-4 UN: 1449 ADR: 5.1,II Merck Index: 14,989 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...