Aluminium isopropoxide 98+%

Supplier: Thermo Scientific Chemicals
Danger

14007-A1 14007-A7
CAAA14007-A1EA 105.54 CAD
CAAA14007-A1 CAAA14007-A7
Aluminium isopropoxide 98+%
Aluminium isopropoxide

Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates


Purity: 98+% CAS No.: 555-31-7 Molecular Formula: Al[OCH(CH3)2]3 Formula Weight: 204.25 Physical Form: Granular Density: 1.035 Refractive Index: 1.0346 Sensitivity: Moisture Sensitive Melting Point: 131-135° Boiling Point: 135°/10mm Solubility: Soluble in alcohol, benzene, and toluene. Decomposes in water Merck Reference: 14,347 UN No.: 3181 MDL No.: MFCD00008870


Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates

Formula: C₉H₂₁AlO₃
Boiling Pt: 135 °C (10 mmHg)
Melting Pt: 131…135 °C
Density: 1.035
Storage Temperature: Ambient
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Merck Index: 14,347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR