Titanium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13999-BP
13999-BS
CAAA13999-BPEA
224.36
CAD
CAAA13999-BP
CAAA13999-BS
Titanium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)
Titanium
MDL: MFCD00011264
Purity: 99.99%
CAS No.: 7440-32-6
Physical Form: ≈5.7g/cm
Odor: Odorless
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...