Ruthenium ≥99.9% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

13994-04 13994-09 13994-18
CAAA13994-04EA 376.04 CAD
CAAA13994-04 CAAA13994-09 CAAA13994-18
Ruthenium ≥99.9% (metals basis), powder -325 mesh
Ruthenium

MDL: MFCD00011207


Purity: 99.9% CAS No.: 7440-18-8 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011207

Formula: Ru
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II
Merck Index: 12,08452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR