Platinum disulfide Pt ≥ 74,8% (≥74.8% Pt)
Supplier: Thermo Scientific Chemicals
13992-03
13992-06
CAAA13992-03EA
576.7
CAD
CAAA13992-03
CAAA13992-06
Platinum disulfide Pt ≥ 74,8% (≥74.8% Pt)
Platinum disulphide
MDL: MFCD00016293
CAS No.: 12038-21-0
Molecular Formula: PtS2
Formula Weight: 259.22
Physical Form: Powder
Melting Point: 225-250° dec.
MDL No.: MFCD00016293
Formula:
PtS₂ Melting Pt: 225…250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016293 CAS Number: 12038-21-0 EINECS: 234-876-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...