Copper ≥99% (metals basis), powder -325 mesh, 10% max +325 mesh

Supplier: Thermo Scientific Chemicals
Warning

13990-18 13990-36 13990-A3 13990-A9
CAAA13990-18EA 79 CAD
CAAA13990-18 CAAA13990-36 CAAA13990-A3 CAAA13990-A9
Copper ≥99% (metals basis), powder -325 mesh, 10% max +325 mesh
Copper

MDL: MFCD00010965


Purity: 99% CAS No.: 7440-50-8 Physical Form: APS 8-11 micron (FSSS) Appearance: Copper Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR