Barium sulfate 97%
Supplier: Thermo Scientific Chemicals
13989-22
13989-A1
13989-A7
CAAA13989-22EA
64.62
CAD
CAAA13989-22
CAAA13989-A1
CAAA13989-A7
Barium sulfate 97%
Barium sulphate
MDL: MFCD00003455
Insoluble in water, dilute acids, alcohol. Soluble in hot, concentrated H2SO4
Purity: 97%
CAS No.: 7727-43-7
Molecular Formula: BaSO4
Formula Weight: 233.40
Physical Form: 1-4 Micron Powder
Density: 4.5
Melting Point: 1580° dec.
Solubility: Insoluble in water, dilute acids, alcohol. Soluble in hot, concentrated H2SO4
Merck Reference: 14,994
MDL No.: MFCD00003455
Formula:
BaSO₄ Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 1580 °C (dec.) Density: 4.5 Storage Temperature: Ambient |
MDL Number:
MFCD00003455 CAS Number: 7727-43-7 EINECS: 231-784-4 Merck Index: 14,994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...