Gallium trifluoride ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13987-03 13987-06
CAAA13987-03EA 225.94 CAD
CAAA13987-03 CAAA13987-06
Gallium trifluoride ≥99.995% (metals basis)
Gallium trifluoride

MDL: MFCD00011021


Purity: 99.999% CAS No.: 7783-51-9 Molecular Formula: GaF3 Formula Weight: 126.72 Physical Form: -20 Mesh Powder Density: 4.47 Appearance: White Odor: Odorless Melting Point: 800° subl. Boiling Point: ca 1000° Merck Reference: 14,4354 UN No.: 3260 MDL No.: MFCD00011021

Formula: F₃Ga
MW: 126.72
Boiling Pt: 1000
Melting Pt: 800 (subl.)
Density: 4.47
Storage Temperature: Ambient
MDL Number: MFCD00011021
CAS Number: 7783-51-9
EINECS: 232-004-5
UN: 3260
ADR: 8 III
Merck Index: 14,4354

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR