Indium ≥99.99% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
13982-FI
13982-GJ
13982-HW
CAAA13982-FIEA
216.46
CAD
CAAA13982-FI
CAAA13982-GJ
CAAA13982-HW
Indium ≥99.99% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Indium
MDL: MFCD00134048
Purity: 99.99%
CAS No.: 7440-74-6
Physical Form: ≈4.6g/50x50mm
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...