Indium phosphide ≥99.9999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indium(III) phosphide, Indium phosphide
13974-03
13974-06
CAAA13974-03EA
347.6
CAD
CAAA13974-03
CAAA13974-06
Indium phosphide ≥99.9999% (metals basis)
Indium phosphide
MDL: MFCD00016153
Purity: 99.9999%
CAS No.: 22398-80-7
Molecular Formula: InP
Formula Weight: 145.79
Physical Form: Pieces
Density: 4.81
Melting Point: 1070°
Merck Reference: 14,4953
MDL No.: MFCD00016153
Formula:
InP MW: 145.79 g/mol Melting Pt: 1070 °C Density: 4.81 Storage Temperature: Ambient |
MDL Number:
MFCD00016153 CAS Number: 22398-80-7 EINECS: 244-959-5 Merck Index: 14,4953 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...