Nickel chromium alloy (80:20 w%), wire, Ø 0.51 mm (0.02 in)

Supplier: Thermo Scientific Chemicals
Warning

13967-H4 13967-H9
CAAA13967-H4EA 186.44 CAD
CAAA13967-H4 CAAA13967-H9
Nickel chromium alloy (80:20 w%), wire, Ø 0.51 mm (0.02 in)
Nickel Chromium alloy (80:20 w%)

MDL: MFCD00673110


CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: ≈1.86g/m Odor: Odorless MDL No.: MFCD00673110

Storage Temperature: Ambient
MDL Number: MFCD00673110
CAS Number: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR