Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)

Supplier: Thermo Scientific Chemicals
Warning

13966-H4 13966-H9
CAAA13966-H4EA 57.2 CAD
CAAA13966-H4 CAAA13966-H9 CAAA13966-J5
Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)
Nickel Chromium alloy (80:20 w%)

High resistivity material for electronics industry


CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: ≈0.14g/m Odor: Odorless MDL No.: MFCD00673110


High resistivity material for electronics industry

Storage Temperature: Ambient
MDL Number: MFCD00673110
CAS Number: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR