Neodymium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

13964-FF 13964-FI
CAAA13964-FFEA 238.58 CAD
CAAA13964-FF CAAA13964-FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.1 mm (0.004 in)
Neodymium

MDL: MFCD00011130


Purity: 99.9% CAS No.: 7440-00-8 Physical Form: ≈0.43g/25x25mm, Packaged in mylar under argon Odor: Odorless UN No.: 3178 MDL No.: MFCD00011130

Formula: Nd
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR