Praseodymium 99,9 REO basis, foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

13962-FF 13962-FI
CAAA13962-FFEA 529.3 CAD
CAAA13962-FF CAAA13962-FI
Praseodymium 99,9 REO basis, foil, Thickness: 0.1 mm (0.004 in)
Praseodymium

MDL: MFCD00011174


Purity: 99.9% CAS No.: 7440-10-0 Physical Form: ≈0.42g/25x25mm, Packaged in mylar under argon Odor: Odorless UN No.: 3178 MDL No.: MFCD00011174

Formula: Pr
MW: 140.91
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR