Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

13960-FF 13960-FI
CAAA13960-FFEA 289.14 CAD
CAAA13960-FF CAAA13960-FI
Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Yttrium

MDL: MFCD00011468


Purity: 99.9% CAS No.: 7440-65-5 Physical Form: ≈0.7g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011468

Formula: Y
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR